Bicyclo[3.3.1]-9-nonanon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bicyclo[3.3.1]nonan-9-one

M-Clarity™ Qualitätsstufe = MQ100

186015-1G
SIAL186015-1GEA 196 EUR
SIAL186015-1G
Bicyclo[3.3.1]-9-nonanon, Sigma-Aldrich®
Bicyclo[3.3.1]-9-nonanon
Formel: C₉H₁₄O
Molecular Weight: 138.21
Schmelzpunkt: 156…158 °C
MDL: MFCD00074752
CAS-Nummer: 17931-55-4
EINECS: 241-868-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR