2'-Aminoacetophenon Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Aminophenyl)ethanone hydrochloride, 2′-Aminoacetophenone hydrochloride, o-Aminoacetophenone hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

186228-25G
SIAL186228-25GEA 207 EUR
SIAL186228-25G
2'-Aminoacetophenon Hydrochlorid, Sigma-Aldrich®
2'-Aminoacetophenon Hydrochlorid
Formel: C₈H₁₀ClNO
Molecular Weight: 171,6241 g/mol
MDL: MFCD00035477
CAS-Nummer: 25384-14-9
EINECS: 246-930-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR