2-Naphthalinmethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

187313-25G 187313-5G
SIAL187313-25GEA 292 EUR
SIAL187313-25G SIAL187313-5G
2-Naphthalinmethanol, Sigma-Aldrich®
2-Naphthalinmethanol
Formel: C₁₁H₁₀O
Molecular Weight: 158.2000 g/mol
MDL: MFCD00004124
CAS-Nummer: 1592-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR