3-Methyl-4-nitrobenzylalkohol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-(Hydroxymethyl)-2-nitrotoluol
M-Clarity™ Qualitätsstufe = MQ100
187631-10G
SIAL187631-10GEA
122
EUR
SIAL187631-10G
3-Methyl-4-nitrobenzylalkohol, Sigma-Aldrich®
3-Methyl-4-nitrobenzylalkohol
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Schmelzpunkt: 60…62,5 °C |
MDL:
MFCD00007171 CAS-Nummer: 80866-75-7 EINECS: 279-578-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...