2,5-Dimethoxybenzylalkohol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
187879-10G
SIAL187879-10GEA
153
EUR
SIAL187879-10G
2,5-Dimethoxybenzylalkohol, Sigma-Aldrich®
2,5-Dimethoxybenzylalkohol
Formel:
C₉H₁₂O₃ Siedepunkt: 122 °C (1013 hPa) |
MDL:
MFCD00004615 CAS-Nummer: 33524-31-1 EINECS: 251-562-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...