5-Phenyl-1-pentanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

188220-5G
SIAL188220-5GEA 127 EUR
SIAL188220-5G
5-Phenyl-1-pentanol, Sigma-Aldrich®
5-Phenyl-1-pentanol
Formel: (C₆H₅)CH₂CH₂CH₂CH₂CH₂OH
Molecular Weight: 164,25 g/mol
Dichte: 0,975 g/cm³ (20 °C)
MDL: MFCD00002979
CAS-Nummer: 10521-91-2
EINECS: 234-064-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR