3-Methyl-2-nitrobenzylalkohol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(Hydroxymethyl)-2-nitrotoluol
M-Clarity™ Qualitätsstufe = MQ100
188263-5G
SIAL188263-5GEA
117
EUR
SIAL188263-5G
3-Methyl-2-nitrobenzylalkohol, Sigma-Aldrich®
3-Methyl-2-nitrobenzylalkohol
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Schmelzpunkt: 45…47 °C |
MDL:
MFCD00007182 CAS-Nummer: 80866-76-8 EINECS: 279-579-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...