4-Chlor-2-nitrobenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

188271-5G
SIAL188271-5GEA 100 EUR
SIAL188271-5G
4-Chlor-2-nitrobenzylalkohol, Sigma-Aldrich®
4-Chlor-2-nitrobenzylalkohol
Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
Schmelzpunkt: 89…90 °C
MDL: MFCD00007217
CAS-Nummer: 22996-18-5
EINECS: 245-374-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR