2-Mercapto-5-methyl-1,3,4-thiadiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-5-thio-1,3,4-thiadiazole, 5-Methyl-1,3,4-thiadiazole-2-thiol

M-Clarity™ Qualitätsstufe = MQ100

188573-25G
SIAL188573-25GEA 27.4 EUR
SIAL188573-25G
2-Mercapto-5-methyl-1,3,4-thiadiazol, Sigma-Aldrich®
2-Mercapto-5-methyl-1,3,4-thiadiazole
Formel: C₃H₄N₂S₂
Molecular Weight: 132,21 g/mol
MDL: MFCD00038349
CAS-Nummer: 29490-19-5
EINECS: 249-667-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR