2,2-Bis(4-chlorphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-DDOH

188883-1G
SIAL188883-1GEA 200 EUR
SIAL188883-1G
2,2-Bis(4-chlorphenyl)ethanol, Sigma-Aldrich®
2,2-Bis(4-chlorphenyl)ethanol
Formel: C₁₄H₁₂Cl₂O
Molecular Weight: 267,15 g/mol
CAS-Nummer: 2642-82-2
EINECS: 220-148-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR