Nitro-tert-butan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Methyl-2-nitropropane
M-Clarity™ Qualitätsstufe = MQ100
189022-25G
SIAL189022-25GEA
139
EUR
SIAL189022-25G
Nitro-tert-butan, Sigma-Aldrich®
Nitro-tert-butan
Formel:
C₄H₉NO₂ Molecular Weight: 103,12 g/mol Siedepunkt: 126 °C (1013 hPa) Dichte: 1,059 g/cm³ (20 °C) |
MDL:
MFCD00007393 CAS-Nummer: 594-70-7 EINECS: 209-851-4 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...