5-Brom-2-methoxybenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (5-Brom-2-methoxyphenyl)methanol, 4-Brom-3-(hydroxymethyl)anisol

189693-1G
SIAL189693-1GEA 30.2 EUR
SIAL189693-1G
5-Brom-2-methoxybenzylalkohol, Sigma-Aldrich®
5-Brom-2-methoxybenzylalkohol
Formel: C₈H₉BrO₂
Molecular Weight: 217,06 g/mol
MDL: MFCD00004630
CAS-Nummer: 80866-82-6
EINECS: 279-586-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR