Chromon-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid

189782-5G
SIAL189782-5GEA 44.3 EUR
SIAL189782-5G
Chromon-2-carbonsäure, Sigma-Aldrich®
Chromon-2-carbonsäure
Formel: C₁₀H₆O₄
Molecular Weight: 190,15 g/mol
MDL: MFCD00006838
CAS-Nummer: 4940-39-0
EINECS: 225-583-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR