Ethylmagnesiumbromid ∼39% (w/w) 3 mol/l in Diethylether, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

189871-800ML 189871-100ML 189871-8L
SIAL189871-800MLEA 336 EUR
SIAL189871-800ML SIAL189871-100ML SIAL189871-8L
Ethylmagnesiumbromid ∼39% (w/w) 3 mol/l in Diethylether, Sigma-Aldrich®
Ethylmagnesium bromide
Formel: C₂H₅BrMg
MDL: MFCD00000043
CAS-Nummer: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR