Pyridiniumchlorochromat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
190144-100G
190144-25G
190144-500G
SIAL190144-100GEA
66.7
EUR
SIAL190144-100G
SIAL190144-25G
SIAL190144-500G
Pyridiniumchlorochromat, Sigma-Aldrich®
Pyridinium chlorochromate
Formel:
C₅H₆ClCrNO₃ Schmelzpunkt: 205 °C |
MDL:
MFCD00013106 CAS-Nummer: 26299-14-9 EINECS: 247-595-5 UN: 1479 ADR: 5.1,II Merck Index: 13,08064 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...