L-Phenylalaninol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-Phenylalaninol, L(-)-Phenylalaninol
M-Clarity™ Qualitätsstufe = MQ100
190438-10G
190438-1G
SIAL190438-10GEA
114
EUR
SIAL190438-10G
SIAL190438-1G
L-Phenylalaninol, Sigma-Aldrich®
L-Phenylalaninol
Formel:
C₉H₁₃NO Siedepunkt: 278 °C (1013 hPa) Schmelzpunkt: 91…94 °C |
MDL:
MFCD00004732 CAS-Nummer: 3182-95-4 EINECS: 221-674-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...