2,4,5-Trimethoxybenzylalkohol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

190489-5G
SIAL190489-5GEA 119 EUR
SIAL190489-5G
2,4,5-Trimethoxybenzylalkohol, Sigma-Aldrich®
2,4,5-Trimethoxybenzylalkohol
Formel: C₁₀H₁₄O₄
Molecular Weight: 198,22 g/mol
MDL: MFCD00021966
CAS-Nummer: 30038-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR