L(+)-Isoleucinol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-Isoleucinol, (S)-(+)-Isoleucinol, (2S,3S)-2-Amino-3-methyl-1-pentanol, (2S,3S)-2-Amino-3-methylpentan-1-ol

M-Clarity™ Qualitätsstufe = MQ100

190527-1G
SIAL190527-1GEA 53 EUR
SIAL190527-1G
L(+)-Isoleucinol, Sigma-Aldrich®
L(+)-Isoleucinol
Formel: C₆H₁₅NO
Molecular Weight: 117.19 g/mol
MDL: MFCD00004731
CAS-Nummer: 24629-25-2
EINECS: 246-371-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR