3-Amino-5-methylthio-1H-1,2,4-triazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-5-(methylmercapto)-1H-1,2,4-triazole

M-Clarity™ Qualitätsstufe = MQ100

190683-25G
SIAL190683-25GEA 146 EUR
SIAL190683-25G
3-Amino-5-methylthio-1H-1,2,4-triazol, Sigma-Aldrich®
3-Amino-5-methylthio-1H-1,2,4-triazol
Formel: C₃H₆N₄S
Molecular Weight: 130,17 g/mol
MDL: MFCD00005232
CAS-Nummer: 45534-08-5
EINECS: 256-242-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR