α-Chlor-3-nitrotoluol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Nitrobenzylchlorid, m-Nitrobenzylchlorid, 1-(Chlormethyl)-3-nitrobenzol, α-Chlor-m-nitrotoluol
191167-10G
191167-50G
SIAL191167-10GEA
60
EUR
SIAL191167-10G
SIAL191167-50G
α-Chlor-3-nitrotoluol, Sigma-Aldrich®
α-Chlor-3-nitrotoluol
Formel:
C₇H₆ClNO₂ |
MDL:
MFCD00007272 CAS-Nummer: 619-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...