4-Ethylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck

191280-25G 191280-5G
SIAL191280-25GEA 77.7 EUR
SIAL191280-25G SIAL191280-5G
4-Ethylbenzoesäure, Sigma-Aldrich®
4-Ethylbenzoesäure
Formel: C₉H₁₀O₂
Molecular Weight: 150.18 g/mol
MDL: MFCD00002570
CAS-Nummer: 619-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR