4-tert-Butyl-α-chlortoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-tert-Butylbenzylchlorid, p-tert-Butyl-α-chlortoluol, p-tert-Butylbenzylchlorid

M-Clarity™ Qualitätsstufe = MQ100

191531-5G
SIAL191531-5GEA 37 EUR
SIAL191531-5G
4-tert-Butyl-α-chlortoluol, Sigma-Aldrich®
4-tert-Butyl-α-chlortoluol
Formel: C₁₁H₁₅Cl
Molecular Weight: 182,69 g/mol
MDL: MFCD00000918
CAS-Nummer: 19692-45-6
EINECS: 243-228-8
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR