3-Methyl-1-phenyl-2-phospholen-1-oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-1-phenyl-2,3-dihydro-1H-phosphole 1-oxide

191744-10G
SIAL191744-10GEA 206 EUR
SIAL191744-10G
3-Methyl-1-phenyl-2-phospholen-1-oxid, Sigma-Aldrich®
3-Methyl-1-phenyl-2-phospholen-1-oxid
Formel: C₁₁H₁₃OP
Molecular Weight: 192,2 g/mol
MDL: MFCD00014518
CAS-Nummer: 707-61-9
EINECS: 211-901-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR