α-Chlor-5-methyl-2-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methyl-2-nitrobenzyl chloride

191779-10G
SIAL191779-10GEA 37.9 EUR
SIAL191779-10G
α-Chlor-5-methyl-2-nitrotoluol, Sigma-Aldrich®
α-Chlor-5-methyl-2-nitrotoluol
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
Schmelzpunkt: 41…43 °C
CAS-Nummer: 66424-91-7
EINECS: 266-359-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR