n-Octyl-β-D-maltosid, Sigma-Aldrich®

Lieferant: Merck


MQ100

19181-1G
SIAL19181-1GEA 307 EUR
SIAL19181-1G
n-Octyl-β-D-maltosid, Sigma-Aldrich®
n-Octyl-β-D-maltosid
Formel: C₂₀H₃₈O₁₁
CAS-Nummer: 82494-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR