cis-Bicyclo[3.3.0]octan-3,7-dion, Sigma-Aldrich®

Lieferant: Merck

Synonyme: cis-Tetrahydropentalene-2,5(1H,3H)-dione

M-Clarity™ Qualitätsstufe = MQ100

192104-1G
SIAL192104-1GEA 154 EUR
SIAL192104-1G
cis-Bicyclo[3.3.0]octan-3,7-dion, Sigma-Aldrich®
cis-Bicyclo[3.3.0]octan-3,7-dion
Formel: C₈H₁₀O₂
Molecular Weight: 138,1600 g/mol
MDL: MFCD00003771
CAS-Nummer: 51716-63-3
EINECS: 257-357-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR