Borwasserstoff-Dimethylsulfid-Komplex 2 mol/l in THF, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
192120-100ML
192120-800ML
SIAL192120-100MLEA
91.6
EUR
SIAL192120-100ML
SIAL192120-800ML
Borwasserstoff-Dimethylsulfid-Komplex 2 mol/l in THF, Sigma-Aldrich®
Borwasserstoff-Dimethylsulfid-Komplex
Formel:
C₂H₉BS |
MDL:
MFCD00013189 CAS-Nummer: 13292-87-0 EINECS: 236-313-6 UN: 3399 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...