Isochinolin-N-oxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Isoquinoline N-Oxide
M-Clarity™ Qualitätsstufe = MQ100
192694-5G
SIAL192694-5GEA
40.6
EUR
SIAL192694-5G
Isochinolin-N-oxid, Sigma-Aldrich®
Isochinolin-N-oxid
Formel:
C₉H₇NO Molecular Weight: 145.16 g/mol |
MDL:
MFCD00006903 CAS-Nummer: 1532-72-5 EINECS: 216-242-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...