Z-N-Me-Aib-OH ≥98%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Carbobenzyl-Oxy-N,2-Dimethylalanine, Z-N-methyl-alpha-aminoisobutyric acid, 2-methyl-2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid, Z-N,2-Dimethylalanine

192902-1G
SIAL192902-1GEA 22.9 EUR
SIAL192902-1G
Z-N-Me-Aib-OH ≥98%, Sigma-Aldrich®
Z-N-Me-Aib-OH
Formel: C₆H₅CH₂OCON(CH₃)C(CH₃)₂CO₂H
Molecular Weight: 251,28 g/mol
Schmelzpunkt: 135…138 °C (Lit.)
MDL: MFCD00191898
CAS-Nummer: 144332-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR