2-Amino-4-(2-methyl-2-propanyl)phenol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
193283-10G
SIAL193283-10GEA
59.8
EUR
SIAL193283-10G
2-Amino-4-(2-methyl-2-propanyl)phenol, Sigma-Aldrich®
2-Amino-4-tert-butylphenol
Formel:
C₁₀H₁₅NO |
MDL:
MFCD00007698 CAS-Nummer: 1199-46-8 EINECS: 214-844-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...