(α,α,α-Trifluor-3-tolyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (3-(Trifluormethyl)phenyl)essigsäure

M-Clarity™ Qualitätsstufe = MQ200

193356-10G
SIAL193356-10GEA 63.2 EUR
SIAL193356-10G
(α,α,α-Trifluor-3-tolyl)essigsäure, Sigma-Aldrich®
(α,α,α-Trifluor-3-tolyl)essigsäure
Formel: (C₆H₄(CF₃))CH₂COOH
Molecular Weight: 204,15 g/mol
MDL: MFCD00004339
CAS-Nummer: 351-35-9
EINECS: 206-511-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR