(S)-3-(Boc-amino)-3-(4-chlorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

19343-500MG-F
SIAL19343-500MG-FEA 258 EUR
SIAL19343-500MG-F
(S)-3-(Boc-amino)-3-(4-chlorphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(4-chlorphenyl)propionsäure
Formel: C₁₄H₁₈ClNO₄
Molecular Weight: 299,7500 g/mol
MDL: MFCD03427922
CAS-Nummer: 479064-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR