α-Chlor-2-iodtoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Iodobenzyl chloride

193496-10G
SIAL193496-10GEA 179 EUR
SIAL193496-10G
α-Chlor-2-iodtoluol, Sigma-Aldrich®
α-Chlor-2-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252,48 g/mol
CAS-Nummer: 59473-45-9
EINECS: 261-779-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR