2,6-Dichlor-3-nitropyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
193585-10G
SIAL193585-10GEA
72.8
EUR
SIAL193585-10G
2,6-Dichlor-3-nitropyridin, Sigma-Aldrich®
2,6-Dichloro-3-nitropyridine
Formel:
C₅H₂Cl₂N₂O₂ Molecular Weight: 192,99 g/mol Schmelzpunkt: 67…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006234 CAS-Nummer: 16013-85-7 EINECS: 240-151-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...