(±)-1-Indanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-1-Hydroxyindan

M-Clarity™ Qualitätsstufe = MQ200

193739-5G
SIAL193739-5GEA 0 EUR
SIAL193739-5G
(±)-1-Indanol, Sigma-Aldrich®
(±)-1-Indanol
Formel: C₉H₁₀O
Molecular Weight: 134,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00003797
CAS-Nummer: 6351-10-6
EINECS: 228-755-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR