2,5-Diaminopyridin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

193755-25G
SIAL193755-25GEA 323 EUR
SIAL193755-25G
2,5-Diaminopyridin Dihydrochlorid, Sigma-Aldrich®
2,5-Diaminopyridin Dihydrochlorid
Formel: C₅H₉Cl₂N₃
Molecular Weight: 182,05 g/mol
Schmelzpunkt: 264 °C
MDL: MFCD00040818
CAS-Nummer: 26878-35-3
EINECS: 248-083-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR