2-(2-Chlorphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

193844-5G
SIAL193844-5GEA 85.9 EUR
SIAL193844-5G
2-(2-Chlorphenyl)ethanol, Sigma-Aldrich®
2-(2-Chlorphenyl)ethanol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD00002888
CAS-Nummer: 19819-95-5
EINECS: 243-348-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR