4'-Chlor-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chloro-3-nitroacetophenone

M-Clarity™ Qualitätsstufe = MQ100

193968-5G
SIAL193968-5GEA 42.7 EUR
SIAL193968-5G
4'-Chlor-3'-nitroacetophenon, Sigma-Aldrich®
4'-Chlor-3'-nitroacetophenon
Formel: C₈H₆ClNO₃
MDL: MFCD00007083
CAS-Nummer: 5465-65-6
EINECS: 226-769-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR