Ethyl-3-piperidincarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

194360-100G 194360-25G
SIAL194360-100GEA 88.7 EUR
SIAL194360-100G SIAL194360-25G
Ethyl-3-piperidincarboxylat, Sigma-Aldrich®
Ethyl-3-piperidincarboxylat
Formel: C₈H₁₅NO₂
MDL: MFCD00005991
CAS-Nummer: 5006-62-2
EINECS: 225-681-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR