α-Chlor-2-methyl-3-nitrotoluol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
195367-5G
SIAL195367-5GEA
144
EUR
SIAL195367-5G
α-Chlor-2-methyl-3-nitrotoluol, Sigma-Aldrich®
α-Chlor-2-methyl-3-nitrotoluol
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,61 g/mol Schmelzpunkt: 43…44 °C |
MDL:
MFCD00007165 CAS-Nummer: 60468-54-4 EINECS: 262-249-3 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...