4-(Methylamino)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

195510-1G
SIAL195510-1GEA 35.8 EUR
SIAL195510-1G
4-(Methylamino)pyridin, Sigma-Aldrich®
4-(Methylamino)pyridine
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
MDL: MFCD00152625
CAS-Nummer: 1121-58-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR