2,3,4-Trimethoxybenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2,3,4-Trimethoxyphenyl)methanol

M-Clarity™ Qualitätsstufe = MQ100

195863-5G
SIAL195863-5GEA 49.4 EUR
SIAL195863-5G
2,3,4-Trimethoxybenzylalkohol, Sigma-Aldrich®
2,3,4-Trimethoxybenzylalkohol
Formel: C₁₀H₁₄O₄
Molecular Weight: 198,22 g/mol
MDL: MFCD00004613
CAS-Nummer: 71989-96-3
EINECS: 276-270-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR