2-Thienylacetylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

M-Clarity™ Qualitätsstufe = MQ200

195995-25G
SIAL195995-25GEA 81.4 EUR
SIAL195995-25G
2-Thienylacetylchlorid, Sigma-Aldrich®
2-Thienylacetylchlorid
Formel: C₆H₅ClOS
Molecular Weight: 160,62 g/mol
MDL: MFCD00005456
CAS-Nummer: 39098-97-0
EINECS: 254-290-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR