α,4-Dichlor-2-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chloro-2-nitrobenzyl chloride

196258-2G
SIAL196258-2GEA 93.6 EUR
SIAL196258-2G
α,4-Dichlor-2-nitrotoluol, Sigma-Aldrich®
α,4-Dichlor-2-nitrotoluol
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
Schmelzpunkt: 37…39 °C
CAS-Nummer: 938-71-6
EINECS: 213-345-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR