2-Phenyl-4-chinolincarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Phenyl-4-quinolinecarboxylic acid, Cinchophen

M-Clarity™ Qualitätsstufe = MQ100

196479-50G
SIAL196479-50GEA 558 EUR
SIAL196479-50G
2-Phenyl-4-chinolincarbonsäure, Sigma-Aldrich®
2-Phenylquinoline-4-carboxylic acid
Formel: C₁₆H₁₁NO₂
Molecular Weight: 249.27 g/mol
MDL: MFCD00006750
CAS-Nummer: 132-60-5
EINECS: 205-067-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR