Tetrafluorphthalonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrafluor-1,2-benzoldicarbonitril, Tetrafluorbenzol-1,2-dicarbonitril, Tetrafluorphthalsäuredinitril, Tetrafluor-1,2-dicyanbenzol, Tetrafluor-1,2-dicyanobenzol

M-Clarity™ Qualitätsstufe = MQ100

196819-1G
SIAL196819-1GEA 27.2 EUR
SIAL196819-1G
Tetrafluorphthalonitril, Sigma-Aldrich®
Tetrafluorphthalonitril
Formel: C₈F₄N₂
Molecular Weight: 200,1 g/mol
MDL: MFCD00001774
CAS-Nummer: 1835-65-0
EINECS: 217-400-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR