4-Morpholinoanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Morpholinyl)aniline

M-Clarity™ Qualitätsstufe = MQ200

197157-5G
SIAL197157-5GEA 37.8 EUR
SIAL197157-5G
4-Morpholinoanilin, Sigma-Aldrich®
4-Morpholinoanilin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,23 g/mol
MDL: MFCD00006169
CAS-Nummer: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR