Methylcyclohexancarboxylat, Sigma-Aldrich®

Lieferant: Merck


MQ100

197424-10G
SIAL197424-10GEA 60.4 EUR
SIAL197424-10G
Methylcyclohexancarboxylat, Sigma-Aldrich®
Methylcyclohexancarboxylat
Formel: C₈H₁₄O₂
Molecular Weight: 142.2 g/mol
MDL: MFCD00001458
CAS-Nummer: 4630-82-4
EINECS: 225-050-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR