Phenyltrimethylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trimethyl(phenyl)silan

M-Clarity™ Qualitätsstufe = MQ100

197734-25G
SIAL197734-25GEA 96.4 EUR
SIAL197734-25G
Phenyltrimethylsilan, Sigma-Aldrich®
Phenyltrimethylsilan
Formel: C₉H₁₄Si
Molecular Weight: 150.3 g/mol
Siedepunkt: 170
Dichte: 0.873
MDL: MFCD00008271
CAS-Nummer: 768-32-1
EINECS: 212-192-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR