2-(4-Chlorphenoxy)isobuttersäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid
M-Clarity™ Qualitätsstufe = MQ200
197777-50G
SIAL197777-50GEA
103
EUR
SIAL197777-50G
2-(4-Chlorphenoxy)isobuttersäure, Sigma-Aldrich®
2-(4-Chlorphenoxy)isobuttersäure
Formel:
C₁₀H₁₁ClO₃ Molecular Weight: 214,65 g/mol |
MDL:
MFCD00004192 CAS-Nummer: 882-09-7 EINECS: 212-925-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...